Vortex Genie® 2 and 2T Mixer, 220 to 230 V
Supplier: Electron Microscopy Sciences
71841-T 71841 71840-AU 71840-BR 71840-EU 71840-SW 71840T-AU 71840T-BR 71840T-EU 71840T-SW
100493-478EA 820.73 USD
100493-478 100491-820 102097-724 102097-726 102097-728 102097-730 102097-736 102097-738 102097-740 102097-742
Vortex Genie® 2 and 2T Mixer, 220 to 230 V
Shakers and Mixers Vortex Mixers
Versatile Vortex-Genie 2 mixes or shakes fluids in any type container with speed and precision.
  • 220 to 230 V model
This Vortex-Genie has unmatched reliability and multi-task versatility
The 2T has timer function, both Touch and Hands-Free mode automatically turns the unit off after set time. Touch Mode: 1 to 60 seconds. Hands-Free (Switch activated Start/Stop) Mode: 1 to 60 minutes, or continuous un-timed mode. Unit automatically stops at set time – in Touch Mode or Hands-Free (switch activated Start/Stop) Mode. Accuracy ±5%. Repeatability ±1%.
Order Now
SPECIFICATIONS
Environmentally Preferable