Mixer Vortex Genie® 2 and 2T, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71840 71840-T
102091-234EA 709.87 USD
102091-234 102097-722 102097-732 102097-734
Mixer Vortex Genie® 2 and 2T, Electron Microscopy Sciences
Shakers and Mixers Vortex Mixers
Versatile Vortex-Genie 2 mixes or shakes fluids in any type container with speed and precision.
This Vortex-Genie has unmatched reliability and multi-task versatility.
The 2T has timer function, both Touch and Hands-Free mode automatically turns the unit off after set time. Touch Mode: 1 to 60 seconds. Hands-Free (Switch activated Start/Stop) Mode: 1 to 60 minutes, or continuous un-timed mode. Unit automatically stops at set time – in Touch Mode or Hands-Free (switch activated Start/Stop) Mode. Accuracy ±5%. Repeatability ±1%.
It features shaking or vortexing action. Variable speedMore Product Information