Grid Staining Matrix System, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71179-01 71179-05 71179-06 71179-07 71179-08
102097-452EA 195.88 USD
102097-452 102097-454 102097-456 102097-458 102097-460
Grid Staining Matrix System, Electron Microscopy Sciences
Staining Boxes
The Matrix system has a simply alpha-numeric identification system. The unit is not solvent or chemically resistant to acids so all stains should be aqueous based only.
  • Stains up to 25 grids at one time or as little as one
The system requires very little stain and you may use different vessels for each stain.
Información de suministro: Each system includes the Matrix Body and 2 each of the Staining Vessels (red and blue).