Oncyte® Nitrocellulose Film Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70335-50A 70335-50N 70335-51A 70335-51N 70335-52A 70335-52N 70335-53A 70335-53N 70335-54A 70335-54N 70335-55A 70335-55N 70335-56A 70335-56N 70335-57A 70335-57N 70335-58A 70335-58N 70335-59A 70335-59N 70335-60A 70335-60N 70335-61A 70335-61N 70335-62A 70335-62N 70335-63A 70335-63N 70335-70A 70335-70N 70335-71A 70335-71N 70335-72A 70335-72N
102096-694PK 789.28 USD
102096-694 102096-696 102096-698 102096-700 102096-702 102096-704 102096-706 102096-708 102096-710 102096-712 102096-714 102096-716 102096-718 102096-720 102096-722 102096-724 102096-726 102096-728 102096-730 102096-732 102096-734 102096-736 102096-738 102096-740 102096-742 102096-744 102096-746 102096-748 102096-750 102096-752 102096-754 102096-756 102096-758 102096-760
Oncyte® Nitrocellulose Film Slides, Electron Microscopy Sciences
Slides Microarray Slides
ONCYTE® is a three-dimensional microarray comprised of a combination of nitrocellulose polymer and proprietary chemistry that provides increased protein affinity and binding
Nitrocellulose film surfaces are manufactured and quality tested to the most exacting standards. To provide an optimal balance between binding capacity and fluorescence sensitivity, two variants of nitrocellulose coatings available:
The ONCYTE® AVID™ nitrocellulose-coated substrates are for applications that require the highest possible protein binding affinity and capacity. Therefore AVID™ slides are characterized byMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable