Cocoon Boxes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64300-3 64300-3A 64300-3B 64300-3D 64300-4 64300-4A 64300-4B 64300-6C 64300-6Y 64300-8 64300-8A 64300-9 64300-9A 64300-9B 64302-10 64302-20 64302-30
102095-028PK 22.71 USD
102095-028 102095-030 102095-032 102095-034 102095-036 102095-038 102095-040 102095-042 102095-044 102095-046 102095-048 102095-050 102095-052 102095-054 102095-058 102095-060 102095-062
Cocoon Boxes, Electron Microscopy Sciences
Boxes Boxes for Vials
The Revolutionary Way for Mailing and Storing Samples
Boxes - Made from polypropylene, envelope shape, available with or without an inner frame. These boxes are the safest way for mailing or storing fragile samples. They are impact and shock resistant With a living-hinge, opaque-clear, reusable and recyclable. The boxes are available in many sizes and styles to fit your needs. Closing Clips and Security Seals are also available as an option.
Vials – are made from polystyrene and they are tough and crystal clear and come with a friction lid. Cocoon Boxes are also available to accept theseMore Product Information