Stand Alone Cooling Chambers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61955-10 61955-20 61955-21 61955-30 61955-40 61955-41 61955-80
102093-862EA 920.83 USD
102093-862 102093-864 102093-866 102093-868 102093-870 102093-872 102093-874
Stand Alone Cooling Chambers, Electron Microscopy Sciences
Coolers Benchtop Coolers
15 & 50 mL Chambers
Ideal for use on hot or cold plates or for temperature sensitive liquid transfers. Anodized aluminum blocks will not rust or corrode. A variety of options are available to accommodatePCR tubes and trays, 1.5 mL tubes, 15 mL and 50 mL conical tubes. A variety of Specialty Chambers are available to keep samples cool and on ice while you work. Freeze these Chambers overnight for added cooling convenience. These Chambers do not fit the CoolSafe system; use separately (*61955-30 not pictured).
15 & 50 mL Chambers
Each Chamber is an anodized aluminum block. ExcellentMore Product Information