Embedding Wells, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61710-18 61710-24 61710-30 61712-34 61713-50 61714-20 61715-24 61716-10
102093-750EA 649.17 USD
102093-750 102093-752 102093-754 102093-758 102093-760 102093-762 102093-764 102093-766
Embedding Wells, Electron Microscopy Sciences
Embedding Cassettes Embedding Rings
This section offers you a simple system of stainless steel well bars, chucks and freezing blocks, kept cold in cryostat, allow you to perform a variety of precision embedding techniques addressing the most difficult embedding tasks with ease and minimum time needed. 20 to 60 seconds needed to freeze (depending on the well size). Multiple samples handled quickly with no monitoring necessary (for placement of heat extractor). Precise flat or on edge embedding, predictable orientation, liquid samples easily embedded and no crush artifact caused by heat extractor weight. Reduce tissue waste withMore Product Information