Microscope Slide Mailers, Solid Cardboard, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71540 71539-01 71539-10 71541 71540-CS 71541-CS
100488-438PK 16.53 USD
100488-438 100489-334 100489-336 102091-220 102097-600 102097-602
Microscope Slide Mailers, Solid Cardboard, Electron Microscopy Sciences
Slide Mailers
The slide mailers are available in sizes to hold one, two, or four 3 x 1" glass slides.
1-Slide Solid Cardboard Slide Mailer is made from 0.050” (1.29 mm) solid cardboard, measures 33/4 x 15/8” when folded. The mailer holds one 3 x 1” glass slide with or without cover glass and has a thumb groove for easy removal. A special cut corner identifies the top of the mailer.
2-Slide Solid Cardboard Slide Mailer is made from .085” (2.17 mm) cardboard and measures 3 x 41/4” when folded. This mailer is unprinted. Upon request the mailer can be printed with warnings for medical specimens. EachMore Product Information