Round Gasket with Rectangular Hole, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
19071422750
10200-078PK 70.12 USD
10200-078
Round Gasket with Rectangular Hole, Bioptechs Inc.®
Seals Gaskets
A round silicone gasket featuring a rectangular 30 mm I.D.
  • Creates user definable flow characteristics
  • Enables near laminar flow
and two perfusion ports. Designed for use in the FCS2® or FCS3® chamber to seal the Microaqueduct slide.
Certificaciones: CE certified.
Información de suministro: Includes 5 gaskets.