Tischler-Kevorkian Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-6887
10198-868EA 1564.8 USD
10198-868
Tischler-Kevorkian Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Sklar's Tischler-Kevorkian Biopsy Punch is an OB/GYN instrument most commonly used in colposcopy procedures to obtain cervical tissue samples for biopsy
This product is straight with a 20.3cm (8") shaft and 3x9.5mm rectangular bite. It features a single lower tooth and a single upper tooth used to grasp difficult to reach tissue. It also has a locking mechansim above the horn on the handle so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal. This reduces the risk of losing the tissue sample as the instrument is withdrawn. This productMore Product Information