Keyes Cutaneous Punch, OR Grade, Sklar
Supplier: Sklar
06-4142 06-4143 06-4144 06-4145 06-4146 06-4147 06-4148
10194-922EA 105.06 USD
10194-922 10194-924 10194-926 10194-928 10194-930 10194-932 10194-934
Keyes Cutaneous Punch, OR Grade, Sklar
Biopsy Punches
Sklar's Keyes Cutaneous Punch is a dermal diagnostic instrument used to remove skin samples for biopsy.
The round sharp end is pushed into the skin and twisted slightly to cut away a small plug of skin. The solid knurled handle provides a gripping surface for the user who controls the depth of the excision. The Keyes punch is often used in gynecological biopsies where a deeper tissue sample is useful for the pathologist.
Información para pedidos: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 1More Product Information