Nail Nipper, OR Grade, Sklar
Supplier: Sklar
97-0474 97-1046 97-1145 97-1056 97-1309 97-1045 97-1055 97-1308 97-1156 97-1148 97-1146
10184-688EA 364.1 USD
10184-688 10192-362 10192-392 10192-436 10192-470 10193-752 10193-856 10194-062 10194-128 10194-206 75818-076
Nail Nipper, OR Grade, Sklar
Cutters
Nail Nipper is used for removing portions of the toenail or the nail plate.
  • Features double-spring, partially cross-serrated
The plier handles with a locking mechanism that is released upon squeezing the handles. This lock is intended to keep the sharp blades closed when the instrument is not being used.