Kirk Bone Mallet, OR Grade, Sklar
Supplier: Sklar
40-5672
10194-004EA 530.4 USD
10194-004
Kirk Bone Mallet, OR Grade, Sklar
Mallets and Hammers
Kirk bone mallet is an orthopedic instrument used in conjuction with chisels, osteotomes and gouges.
Mallets offer an added swinging force
Censitrac Ready products are industry standard and constructed of stainless steel.
The Kirk has a wide surface area on the head and wide oval handle like the Crane mallet. The Kirk has a solid, heavy head to resist deformation from the impact with other tools.
Información para pedidos: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days,More Product Information