5-Aminoisoindolin-1-one 95

Supplier: Matrix Scientific

Synonyms: 5-aminoisoindolin-1-one

068864-1G
101937-998EA 1629.23 USD
101937-998
5-Aminoisoindolin-1-one 95
5-Aminoisoindolin-1-one

Hazard: Irritant

Formula: C₈H₈N₂O
MW: 148.1600 g/mol
MDL Number: MFCD10000828
CAS Number: 222036-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR