1-Methyl-2-(3-pyridin-3-ylpropyl)piperidine-2-carboxylic acid dihydrochloride

Supplier: Matrix Scientific

068258-250MG
101935-634EA 558.46 USD
101935-634
1-Methyl-2-(3-pyridin-3-ylpropyl)piperidine-2-carboxylic acid dihydrochloride
1-Methyl-2-(3-pyridin-3-ylpropyl)piperidine-2-carboxylic acid dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄Cl₂N₂O₂
MDL Number: MFCD21606196
CAS Number: 1361118-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR