3-(6-Methyl-1',2',3',4',5',6'-hexahydro[2,4']bipyridinyl-4-yl)propionic acid ethyl ester dihydrochloride

Supplier: Matrix Scientific

067470-250MG
101934-058EA 638.46 USD
101934-058
3-(6-Methyl-1',2',3',4',5',6'-hexahydro[2,4']bipyridinyl-4-yl)propionic acid ethyl ester dihydrochloride
3-(6-Methyl-1',2',3',4',5',6'-hexahydro[2,4']bipyridinyl-4-yl)propionic acid ethyl ester dihydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆Cl₂N₂O₂
MDL Number: MFCD21605806
CAS Number: 1361112-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR