N-Cyclopentyl-3-(5,7-dimethyl-2-pyrrolidin-3-ylpyrazolo[1,5-a]pyrimidin-6-yl)propionamide dihydrochloride

Supplier: Matrix Scientific

067338-250MG
101933-794EA 638.46 USD
101933-794
N-Cyclopentyl-3-(5,7-dimethyl-2-pyrrolidin-3-ylpyrazolo[1,5-a]pyrimidin-6-yl)propionamide dihydrochloride
N-Cyclopentyl-3-(5,7-dimethyl-2-pyrrolidin-3-ylpyrazolo[1,5-a]pyrimidin-6-yl)propionamide dihydrochloride

Hazard: Irritant

Formula: C₂₀H₃₁Cl₂N₅O
MW: 428.41 g/mol
MDL Number: MFCD21605779
CAS Number: 1361112-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR