Dimethyl-2,6-dimethyl-4-(4-methylphenyl)-1,4-dihydropyridine-3,5-dicarboxylate

Supplier: Matrix Scientific

066350-500MG
101931-478EA 243.08 USD
101931-478
Dimethyl-2,6-dimethyl-4-(4-methylphenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Dimethyl-2,6-dimethyl-4-(4-methylphenyl)-1,4-dihydropyridine-3,5-dicarboxylate

Hazard: Irritant

Formula: C₁₈H₂₁NO₄
MDL Number: MFCD00610976
CAS Number: 73257-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR