1-tert-Butyl-4-methyl-4-propylpiperidine-1,4-dicarboxylate

Supplier: Matrix Scientific

065781-500MG
101930-344EA 243.08 USD
101930-344
1-tert-Butyl-4-methyl-4-propylpiperidine-1,4-dicarboxylate
1-tert-Butyl-4-methyl-4-propylpiperidine-1,4-dicarboxylate

Hazard: Irritant

Formula: C₁₅H₂₇NO₄
MDL Number: MFCD19103491
CAS Number: 1306738-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR