2-(3-Chloropropanoyl)-1,2,3,4-tetrahydroisoquinoline

Supplier: Matrix Scientific

065762-500MG
101930-306EA 146.15 USD
101930-306
2-(3-Chloropropanoyl)-1,2,3,4-tetrahydroisoquinoline
2-(3-Chloropropanoyl)-1,2,3,4-tetrahydroisoquinoline

Hazard: Irritant

Formula: C₁₂H₁₄ClNO
MW: 223.7 g/mol
MDL Number: MFCD02973833
CAS Number: 10579-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR