2-[(3-Chlorophenyl)amino]butanohydrazide

Supplier: Matrix Scientific

065673-500MG
101930-148EA 243.08 USD
101930-148
2-[(3-Chlorophenyl)amino]butanohydrazide
2-[(3-Chlorophenyl)amino]butanohydrazide

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃O
MW: 227.7 g/mol
MDL Number: MFCD19103408
CAS Number: 1306738-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR