[(2-Hydroxyphenyl)amino](oxo)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-((2-Hydroxyphenyl)amino)-2-oxoacetic acid

065647-500MG
101930-096EA 193.85 USD
101930-096
[(2-Hydroxyphenyl)amino](oxo)acetic acid
[(2-Hydroxyphenyl)amino](oxo)acetic acid

Hazard: Irritant

Formula: C₈H₇NO₄
MW: 181.15 g/mol
MDL Number: MFCD15205424
CAS Number: 89942-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR