5-Amino-N-cyclopropyl-1H-1,2,4-triazole-3-carboxamide

Supplier: Matrix Scientific

065503-500MG
101929-806EA 606.15 USD
101929-806
5-Amino-N-cyclopropyl-1H-1,2,4-triazole-3-carboxamide
5-Amino-N-cyclopropyl-1H-1,2,4-triazole-3-carboxamide

Hazard: Irritant

Formula: C₆H₉N₅O
MDL Number: MFCD17221103
CAS Number: 1279219-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR