1-(2-Chloro-5-nitrophenyl)acetone

Supplier: Matrix Scientific

065463-500MG
101929-726EA 364.62 USD
101929-726
1-(2-Chloro-5-nitrophenyl)acetone
1-(2-Chloro-5-nitrophenyl)acetone

Hazard: Irritant

Formula: C₉H₈ClNO₃
MDL Number: MFCD01861430
CAS Number: 320576-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR