4-Amino-1-methylpiperidine-4-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 4-Amino-1-methyl-4-piperidinecarboxylic acid

065308-500MG
101929-416EA 606.15 USD
101929-416
4-Amino-1-methylpiperidine-4-carboxylic acid
4-Amino-1-methylpiperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₇H₁₄N₂O₂
MW: 158.2 g/mol
MDL Number: MFCD00154385
CAS Number: 15580-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR