3-[6-(Benzyloxy)-1H-indol-1-yl]propanoic acid

Supplier: Matrix Scientific

065088-500MG
101928-976EA 221.54 USD
101928-976
3-[6-(Benzyloxy)-1H-indol-1-yl]propanoic acid
3-[6-(Benzyloxy)-1H-indol-1-yl]propanoic acid

Hazard: Irritant

Formula: C₁₈H₁₇NO₃
MDL Number: MFCD16090140
CAS Number: 1219551-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR