1-Methyl-7-(trifluoromethyl)-1H-indole-2,3-dione

Supplier: Matrix Scientific

064595-500MG
101927-996EA 221.54 USD
101927-996
1-Methyl-7-(trifluoromethyl)-1H-indole-2,3-dione
1-Methyl-7-(trifluoromethyl)-1H-indole-2,3-dione

Hazard: Irritant

Formula: C₁₀H₆F₃NO₂
MDL Number: MFCD09028306
CAS Number: 136622-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR