1-(2-Fluoroethyl)cyclobutanecarbonitrile

Supplier: Matrix Scientific

064410-500MG
101927-644EA 552.31 USD
101927-644
1-(2-Fluoroethyl)cyclobutanecarbonitrile
1-(2-Fluoroethyl)cyclobutanecarbonitrile

Hazard: Irritant

Formula: C₇H₁₀FN
MDL Number: MFCD18064591
CAS Number: 1255147-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR