2-(2-Methoxy-4-methylphenoxy)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

064271-500MG
101927-394EA 364.62 USD
101927-394
2-(2-Methoxy-4-methylphenoxy)quinoline-4-carboxylic acid
2-(2-Methoxy-4-methylphenoxy)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₅NO₄
MDL Number: MFCD18064547
CAS Number: 1255147-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR