4-Amino-5-(5-bromopyridin-3-yl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

064118-500MG
101927-088EA 364.62 USD
101927-088
4-Amino-5-(5-bromopyridin-3-yl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(5-bromopyridin-3-yl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₇H₆BrN₅S
MDL Number: MFCD11202841
CAS Number: 1019061-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR