4-Amino-5-(2,4-dichlorophenyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

064114-500MG
101927-080EA 243.08 USD
101927-080
4-Amino-5-(2,4-dichlorophenyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(2,4-dichlorophenyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₈H₆Cl₂N₄S
MW: 261.13 g/mol
MDL Number: MFCD00269269
CAS Number: 93677-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR