(2-Fluoropyridin-4-yl)methanol ≥97%

Supplier: Matrix Scientific

Synonyms: (2-fluoro-pyridin-4-yl)-methanol, 2-Fluoro-4-hydroxymethylpyridine

064055-1G
101926-900EA 106.15 USD
101926-900
(2-Fluoropyridin-4-yl)methanol ≥97%
(2-Fluoropyridin-4-yl)methanol

Hazard: Irritant

Formula: C₆H₆FNO
MW: 127.12 g/mol
MDL Number: MFCD06200782
CAS Number: 131747-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR