3-[(4-Fluoro-2-nitrophenyl)amino]-5,5-dimethylcyclohex-2-en-1-one

Supplier: Matrix Scientific

063531-500MG
101925-900EA 484.62 USD
101925-900
3-[(4-Fluoro-2-nitrophenyl)amino]-5,5-dimethylcyclohex-2-en-1-one
3-[(4-Fluoro-2-nitrophenyl)amino]-5,5-dimethylcyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₄H₁₅FN₂O₃
MW: 278.29 g/mol
MDL Number: MFCD00656354
CAS Number: 330974-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR