3-[(4-Iodophenyl)amino]cyclohex-2-en-1-one

Supplier: Matrix Scientific

063518-500MG
101925-874EA 484.62 USD
101925-874
3-[(4-Iodophenyl)amino]cyclohex-2-en-1-one
3-[(4-Iodophenyl)amino]cyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₂H₁₂INO
MW: 313.14 g/mol
MDL Number: MFCD02707431
CAS Number: 303019-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR