Methyl-4-methyl-4-nitropentanoate

Supplier: Matrix Scientific

Synonyms: Methyl 4-methyl-4-nitropentanoate

063397-500MG
101925-630EA 243.08 USD
101925-630
Methyl-4-methyl-4-nitropentanoate
Methyl-4-methyl-4-nitropentanoate

Hazard: Irritant

Formula: C₇H₁₃NO₄
MW: 175.19 g/mol
MDL Number: MFCD00047757
CAS Number: 16507-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR