5-Chloro-2-[(4-chlorobenzyl)oxy]benzoyl chloride

Supplier: Matrix Scientific

062133-500MG
101923-358EA 364.62 USD
101923-358
5-Chloro-2-[(4-chlorobenzyl)oxy]benzoyl chloride
5-Chloro-2-[(4-chlorobenzyl)oxy]benzoyl chloride

Hazard: Irritant

Formula: C₁₄H₉Cl₃O₂
MDL Number: MFCD12198007
CAS Number: 1160260-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR