2-(2-Nitrophenoxy)propanoyl chloride

Supplier: Matrix Scientific

061872-500MG
101922-836EA 364.62 USD
101922-836
2-(2-Nitrophenoxy)propanoyl chloride
2-(2-Nitrophenoxy)propanoyl chloride

Hazard: Irritant

Formula: C₉H₈ClNO₄
MW: 229.62 g/mol
MDL Number: MFCD03421505
CAS Number: 360051-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR