N-(3-Methylbenzyl)cyclopropanamine hydrochloride

Supplier: Matrix Scientific

060743-500MG
101920-598EA 364.62 USD
101920-598
N-(3-Methylbenzyl)cyclopropanamine hydrochloride
N-(3-Methylbenzyl)cyclopropanamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClN
MDL Number: MFCD18071313
CAS Number: 1332530-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR