GenePix 4300 and 4400 Microarray Scanner, Molecular Devices
Supplier: Molecular Devices
GENEPIX4400 GENEPIXPROV7 GENEPIX4300
10192-206EA 225621.42 USD
10192-206 10192-216 10192-224
GenePix 4300 and 4400 Microarray Scanner, Molecular Devices
Photodocumentation Systems
The GenePix® 4300A and GenePix® 4400A Scanners from Molecular Devices offer maximum imaging quality with optimal resolution in highly configurable platforms.
  • High-resolution for high-density microarrays
  • Full flexibility in choosing dyes
  • Specified and tested for field uniformity
  • Less noise in your data
  • Less breakable components
  • Designed for high-duty cycle usage
Configurations include 5μm or 2.5μm per-pixel maximum scanning resolution, choices of up to four lasers for excitation, and sixteen emission-wavelength filters
While the GenePix® 43More Product Information