Ethyl-3-amino-6,7-dihydro-5H-cyclopenta[b]thieno[3,2-e]pyridine-2-carboxylate

Supplier: Matrix Scientific

059753-500MG
101919-016EA 306.15 USD
101919-016
Ethyl-3-amino-6,7-dihydro-5H-cyclopenta[b]thieno[3,2-e]pyridine-2-carboxylate
Ethyl-3-amino-6,7-dihydro-5H-cyclopenta[b]thieno[3,2-e]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₂S
MDL Number: MFCD00441106
CAS Number: 166113-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR