5-Chloro-4-iodo-2,3-dimethoxypyridine

Supplier: Matrix Scientific

059500-250MG
101918-278EA 643.08 USD
101918-278
5-Chloro-4-iodo-2,3-dimethoxypyridine
5-Chloro-4-iodo-2,3-dimethoxypyridine

Hazard: Irritant

Formula: C₇H₇ClINO₂
MDL Number: MFCD18803487
CAS Number: 1299607-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR