8-Cyclohexyl-5,6-dihydro-4H-pyrazino[3,2,1-jk]carbazole

Supplier: Matrix Scientific

058449-500MG
101914-830EA 233.85 USD
101914-830
8-Cyclohexyl-5,6-dihydro-4H-pyrazino[3,2,1-jk]carbazole
8-Cyclohexyl-5,6-dihydro-4H-pyrazino[3,2,1-jk]carbazole

Hazard: Irritant

Formula: C₂₀H₂₂N₂
MW: 290.41 g/mol
MDL Number: MFCD00409221
CAS Number: 157056-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR