3-Bromo-5-methylpyrazin-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-3-bromo-5-methylpyrazine

057892-1G
101913-686EA 538.46 USD
101913-686
3-Bromo-5-methylpyrazin-2-amine ≥95%
3-Bromo-5-methylpyrazin-2-amine

Hazard: Irritant

Formula: C5H6BrN3
MW: 188.0300 g/mol
MDL Number: MFCD09750037
CAS Number: 74290-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR