4-Methoxypyridin-3-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-4-methoxypyridine

057361-1G
101913-042EA 510.77 USD
101913-042
4-Methoxypyridin-3-ol ≥95%
4-Methoxypyridin-3-ol

Hazard: Irritant

Formula: C₆H₇NO₂
MW: 125.13 g/mol
MDL Number: MFCD11044354
CAS Number: 153199-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR