1-Benzyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Benzyl 3-ethyl 4-oxopyrrolidine-1,3-dicarboxylate

057291-5G
101912-916EA 438.46 USD
101912-916
1-Benzyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate ≥95%
1-Benzyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₁₅H₁₇NO₅
MW: 291.31 g/mol
MDL Number: MFCD09878816
CAS Number: 51814-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR