2-Thiopheneacrylic acid

Supplier: Matrix Scientific

Synonyms: (E)-3-(Thiophen-2-yl)acrylic acid, 3-(2-Thienyl)acrylic Acid

057190-25G
101912-696EA 100 USD
101912-696
2-Thiopheneacrylic acid
2-Thiopheneacrylic acid

Hazard: Irritant

Formula: C₇H₆O₂S
MW: 154.18 g/mol
Melting Pt: 145…148 °C
MDL Number: MFCD00005455
CAS Number: 15690-25-2

Order Now

Specification Test Results

Melting point 145-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR