3-(4-Biphenyl)propionic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-([1,1'-Biphenyl]-4-yl)propanoic acid

056848-5G
101911-526EA 266.15 USD
101911-526
3-(4-Biphenyl)propionic acid ≥98%
3-(4-Biphenyl)propionic acid

Hazard: Irritant

Formula: C₁₅H₁₄O₂
MW: 226.275 g/mol
Melting Pt: 149…152 °C
MDL Number: MFCD06823979
CAS Number: 35888-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR