1-(2,4,5-Trimethylphenyl)-2-thiourea

Supplier: Matrix Scientific

056835-5G
101911-498EA 178.46 USD
101911-498
1-(2,4,5-Trimethylphenyl)-2-thiourea
1-(2,4,5-Trimethylphenyl)-2-thiourea

Hazard: Irritant

Formula: C₁₀H₁₄N₂S
Melting Pt: 160…162 °C
MDL Number: MFCD00060438
CAS Number: 117174-87-5

Order Now

Specification Test Results

Melting point 160-162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR