4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfinyl}aniline ≥95%

Supplier: Matrix Scientific

054877-500MG
101908-886EA 1109.23 USD
101908-886
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfinyl}aniline ≥95%
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfinyl}aniline

Hazard: Irritant

Formula: C₁₂H₈ClF₃N₂OS
Melting Pt: 120…122 °C
MDL Number: MFCD16140341
CAS Number: 1221792-80-8

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR