Ethyl-4-(benzylamino)-1-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate ≥95%

Supplier: Matrix Scientific

054551-500MG
101907-738EA 921.54 USD
101907-738
Ethyl-4-(benzylamino)-1-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate ≥95%
Ethyl-4-(benzylamino)-1-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₈N₄O₂
MW: 310.36 g/mol
Melting Pt: 144…145 °C
MDL Number: MFCD14584838
CAS Number: 160034-51-5

Order Now

Specification Test Results

Melting point 144-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR